siRNA Details
| Gene Symbol: Ccdc144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178418.4 | NP_848505.2 | 10 | 1502 | ||
| GenBank mRNA | AK133178.1 | - | 8 | 3345 | ||
| BC050811.1 | - | 10 | 1504 | 6772771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc144b| Gene Symbol: Ccdc144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178418.4 | NP_848505.2 | 10 | 1502 | ||
| GenBank mRNA | AK133178.1 | - | 8 | 3345 | ||
| BC050811.1 | - | 10 | 1504 | 6772771 | ||