siRNA Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172862.3 | NP_766450.2 | 1 | 4465 | ||
| GenBank mRNA | AB236662.1 | - | 1 | 4465 | ||
| AJ833643.1 | - | 1 | 4194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frem2| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172862.3 | NP_766450.2 | 1 | 4465 | ||
| GenBank mRNA | AB236662.1 | - | 1 | 4465 | ||
| AJ833643.1 | - | 1 | 4194 | |||