siRNA Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172862.3 | NP_766450.2 | 17 | 8263 | ||
| GenBank mRNA | AB236662.1 | - | 17 | 8263 | ||
| AJ833643.1 | - | 17 | 7992 | |||
| AK031494.1 | - | 7 | 1165 | |||
| BC068185.1 | - | 7 | 1165 | |||

Frem2
Genomic Map