siRNA Details
| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178726.3 | NP_848841.2 | 1 | 980 | ||
| GenBank mRNA | AK032529.1 | - | 1 | 1038 | ||
| AK035912.1 | - | 1 | 983 | |||
| AK045724.1 | - | 1 | 1021 | |||
| AK131646.1 | - | 1 | 967 | |||
| AK142789.1 | - | 1 | 998 | |||
| AK147389.1 | - | 1 | 983 | |||
| AK147876.1 | - | 1 | 1017 | |||
| AK165160.1 | - | 1 | 1013 | |||
| AK220523.1 | - | 1 | 985 | |||
| AY184801.1 | - | 1 | 568 | |||
| BC096031.1 | - | 1 | 980 | 6837066 | ||

Ppm1l
Genomic Map