siRNA Details
| Gene Symbol: Mab21l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172295.4 | NP_758499.3 | 3, 4 | 708 | ||
| GenBank mRNA | AK029040.1 | - | 3, 4 | 447 | ||
| AK161057.1 | - | 3, 4 | 625 | |||
| AK166928.1 | - | 458 | ||||
| AK172389.1 | - | 3, 4 | 657 | |||
| BC037703.1 | - | 3, 4 | 645 | 4166596 | ||

Mab21l3
Genomic Map