siRNA Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033350.2 | NP_001028522.2 | 14, 15 | 2344 | ||
| GenBank mRNA | AK041039.1 | - | 5, 6 | 362 | ||
| AK148665.1 | - | 3, 4 | 139 | |||
| AK156201.1 | - | 14, 15 | 2354 | |||
| AY178734.1 | - | 14, 15 | 2364 | |||
| BC116328.1 | - | 4, 5 | 427 | |||
| BE848854.1 | - | 5, 6 | 498 | |||
| EU051378.1 | - | 13, 14 | 1945 | |||

Bank1
Genomic Map