siRNA Details
| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181989.1 | NP_871789.1 | 2 | 355 | ||
| GenBank mRNA | AK136508.1 | - | 1 | 326 | ||
| AY277588.1 | - | 2 | 355 | |||
| BC104134.1 | - | 2 | 274 | 40045268 | ||
| BC104135.1 | - | 2 | 274 | 40045269 | ||

Sdr16c5
Genomic Map