siRNA Details
| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013526.1 | NP_038554.1 | 1 | 426 | ||
| GenBank mRNA | AJ537425.1 | - | 1 | 426 | ||
| BC141339.1 | - | 1 | 340 | 9055951 | ||
| BC141340.1 | - | 1 | 340 | 9055952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf6| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013526.1 | NP_038554.1 | 1 | 426 | ||
| GenBank mRNA | AJ537425.1 | - | 1 | 426 | ||
| BC141339.1 | - | 1 | 340 | 9055951 | ||
| BC141340.1 | - | 1 | 340 | 9055952 | ||