siRNA Details
| Gene Symbol: Rgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172866.3 | NP_766454.1 | 5, 6 | 773 | ||
| GenBank mRNA | AK089288.1 | - | 5, 6 | 556 | ||
| AK129103.1 | - | 5, 6 | 559 | |||
| AK170117.1 | - | 5, 6 | 570 | |||
| BC086614.1 | - | 5, 6 | 579 | 30533256 | ||

Rgp1
Genomic Map