siRNA Details
| Gene Symbol: Fam221b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175517.3 | NP_780726.1 | 5 | 1416 | ||
| GenBank mRNA | AK076646.1 | - | 5 | 1416 | ||
| BC119804.1 | - | 5 | 1409 | 40107542 | ||
| BC122881.1 | - | 5 | 1409 | 40107552 | ||

Fam221b
Genomic Map