siRNA Details
| Gene Symbol: D630039A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178727.2 | NP_848842.1 | 2 | 710 | ||
| GenBank mRNA | AK033446.1 | - | 2 | 214 | ||
| AK035369.1 | - | 2 | 744 | |||
| AK085537.1 | - | 2 | 691 | |||
| BC030929.1 | - | 2 | 662 | 4911378 | ||
| BC095953.1 | - | 2 | 710 | 5043405 | ||

D630039A03Rik
Genomic Map