siRNA Details
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017427.1 | NP_001017427.1 | 15, 16 | 1783 | ||
GenBank mRNA | AK132691.1 | - | 15, 16 | 1763 | ||
BC030882.1 | - | 11, 12 | 1176 | |||
BC040766.1 | - | 11, 12 | 1165 | |||
BC125539.1 | - | 14, 15 | 1597 | 40130054 | ||
BC137774.1 | - | 14, 15 | 1597 | 8860793 | ||
BC144901.1 | - | 14, 15 | 1533 |
