siRNA Details
| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017427.1 | NP_001017427.1 | 13, 14 | 1591 | ||
| GenBank mRNA | AK132691.1 | - | 13, 14 | 1571 | ||
| BC030882.1 | - | 9, 10 | 984 | |||
| BC040766.1 | - | 9, 10 | 973 | |||
| BC125539.1 | - | 12, 13 | 1405 | 40130054 | ||
| BC137774.1 | - | 12, 13 | 1405 | 8860793 | ||
| BC144901.1 | - | 12, 13 | 1341 | |||

Rasef
Genomic Map