siRNA Details
| Gene Symbol: Bnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172870.4 | NP_766458.3 | 5 | 1018 | ||
| GenBank mRNA | AK030323.1 | - | 5 | 1018 | ||
| AY438375.1 | - | 5 | 993 | |||
| BC110483.2 | - | 4 | 898 | 40048720 | ||
| BC110484.1 | - | 4 | 898 | 40048722 | ||

Bnc2
Genomic Map