siRNA Details
| Gene Symbol: Cyp2j12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100182.2 | NP_001093652.1 | 2 | 361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2j12| Gene Symbol: Cyp2j12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100182.2 | NP_001093652.1 | 2 | 361 | ||