siRNA Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146254.4 | NP_666366.4 | 11, 12 | 1753 | ||
| GenBank mRNA | AK030997.1 | - | 4, 5 | 3438 | ||
| BC028975.1 | - | 11, 12 | 1678 | 5363117 | ||
| BC034130.1 | - | 3, 4 | 733 | 4920514 | ||

Wdr78
Genomic Map