siRNA Details
| Gene Symbol: Dmrta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172296.2 | NP_758500.2 | 3 | 2329 | ||
| GenBank mRNA | AK080774.1 | - | 4 | 2332 | ||
| AY145837.1 | - | 3 | 2301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrta2| Gene Symbol: Dmrta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172296.2 | NP_758500.2 | 3 | 2329 | ||
| GenBank mRNA | AK080774.1 | - | 4 | 2332 | ||
| AY145837.1 | - | 3 | 2301 | |||