siRNA Details
| Gene Symbol: Hpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146256.3 | NP_666368.1 | 1 | 626 | ||
| GenBank mRNA | AK043902.1 | - | 1 | 607 | ||
| AK049217.1 | - | 1 | 606 | |||
| AK078929.1 | - | 1 | 627 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpdl| Gene Symbol: Hpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146256.3 | NP_666368.1 | 1 | 626 | ||
| GenBank mRNA | AK043902.1 | - | 1 | 607 | ||
| AK049217.1 | - | 1 | 606 | |||
| AK078929.1 | - | 1 | 627 | |||