siRNA Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153106.2 | NP_694746.2 | 13 | 1564 | ||
| GenBank mRNA | AF529423.1 | - | 13 | 1540 | ||
| AK139616.1 | - | 13 | 1565 | |||
| BC053724.1 | - | 13 | 1562 | 30051727 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi6| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153106.2 | NP_694746.2 | 13 | 1564 | ||
| GenBank mRNA | AF529423.1 | - | 13 | 1540 | ||
| AK139616.1 | - | 13 | 1565 | |||
| BC053724.1 | - | 13 | 1562 | 30051727 | ||