siRNA Details
| Gene Symbol: Zfp933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198619.2 | NP_941021.1 | 4 | 471 | ||
| GenBank mRNA | AK131865.1 | - | 5 | 495 | ||
| BC058101.1 | - | 4 | 462 | 6822264 | ||
| CB525762.1 | - | 4 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp933| Gene Symbol: Zfp933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198619.2 | NP_941021.1 | 4 | 471 | ||
| GenBank mRNA | AK131865.1 | - | 5 | 495 | ||
| BC058101.1 | - | 4 | 462 | 6822264 | ||
| CB525762.1 | - | 4 | 471 | |||