siRNA Details
Gene Symbol: Klhl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033352.3 | NP_001028524.1 | 4 | 2165 | ||
GenBank mRNA | AK052203.1 | - | 4 | 2140 | ||
AK154493.1 | - | 4 | 2166 | |||
AK158141.1 | - | 4 | 2141 | |||
AK177452.1 | - | 1 | 371 | |||
AK190174.1 | - | 1 | 371 | |||
AK205632.1 | - | 1 | 372 | |||
AK208409.1 | - | 1 | 328 | |||
BC060201.1 | - | 4 | 2154 | |||
BC141158.1 | - | 4 | 2139 | 9055770 | ||
BC141159.1 | - | 4 | 2139 | 9055771 |
