siRNA Details
| Gene Symbol: Ttc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172878.3 | NP_766466.3 | 2 | 282 | ||
| GenBank mRNA | AK030055.1 | - | 2 | 256 | ||
| BC068299.1 | - | 134 | 30094917 | |||
| CJ183638.1 | - | 2 | 285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc34| Gene Symbol: Ttc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172878.3 | NP_766466.3 | 2 | 282 | ||
| GenBank mRNA | AK030055.1 | - | 2 | 256 | ||
| BC068299.1 | - | 134 | 30094917 | |||
| CJ183638.1 | - | 2 | 285 | |||