siRNA Details
| Gene Symbol: Rsbn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080977.1 | NP_001074446.1 | 8 | 2040 | ||
| GenBank mRNA | AK033374.1 | - | 7 | 1808 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsbn1l| Gene Symbol: Rsbn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080977.1 | NP_001074446.1 | 8 | 2040 | ||
| GenBank mRNA | AK033374.1 | - | 7 | 1808 | ||