siRNA Details
| Gene Symbol: Ccdc149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256059.1 | NP_001242988.1 | 12 | 2698 | ||
| GenBank mRNA | AK049058.1 | - | 1 | 278 | ||
| BC025881.1 | - | 1 | 372 | 5027889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc149| Gene Symbol: Ccdc149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256059.1 | NP_001242988.1 | 12 | 2698 | ||
| GenBank mRNA | AK049058.1 | - | 1 | 278 | ||
| BC025881.1 | - | 1 | 372 | 5027889 | ||