siRNA Details
| Gene Symbol: Ugt2b35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172881.3 | NP_766469.1 | 1 | 283 | ||
| GenBank mRNA | AK083294.1 | - | 1 | 283 | ||
| AK149501.1 | - | 1 | 270 | |||
| BC113789.1 | - | 1 | 273 | 40061872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2b35| Gene Symbol: Ugt2b35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172881.3 | NP_766469.1 | 1 | 283 | ||
| GenBank mRNA | AK083294.1 | - | 1 | 283 | ||
| AK149501.1 | - | 1 | 270 | |||
| BC113789.1 | - | 1 | 273 | 40061872 | ||