siRNA Details
| Gene Symbol: Hsd17b13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163486.1 | NP_001156958.1 | 6 | 855 | ||
| NM_198030.2 | NP_932147.2 | 6 | 855 | |||
| GenBank mRNA | AY101768.1 | - | 6 | 782 | ||
| BC019427.1 | - | 6 | 831 | 5132342 | ||
| DV660487.1 | - | 3 | 281 | |||

Hsd17b13
Genomic Map