siRNA Details
Gene Symbol: 2900026A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172884.3 | NP_766472.1 | 6 | 1040 | ||
GenBank mRNA | AK041243.1 | - | 6 | 1019 | ||
AK154484.1 | - | 6 | 1079 | |||
AK171617.1 | - | 6 | 1040 | |||
AK220411.1 | - | 4 | 486 | |||
BC147338.1 | - | 6 | 919 | 9056390 | ||
BC147339.1 | - | 6 | 919 | 9056416 |
