siRNA Details
Gene Symbol: Adgrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081342.1 | NP_001074811.1 | 12 | 1716 | ||
GenBank mRNA | AK153762.1 | - | 12 | 1718 | ||
BC151161.1 | - | 12 | 1418 | 9088050 | ||
BC157994.1 | - | 11 | 1322 | 9007307 | ||
BC158001.1 | - | 11 | 1322 | 9007310 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
