siRNA Details
| Gene Symbol: Slc29a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146257.2 | NP_666369.1 | 7 | 974 | ||
| GenBank mRNA | BC025599.1 | - | 7 | 918 | 5250967 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc29a4| Gene Symbol: Slc29a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146257.2 | NP_666369.1 | 7 | 974 | ||
| GenBank mRNA | BC025599.1 | - | 7 | 918 | 5250967 | |