siRNA Details
| Gene Symbol: Tmem130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177735.4 | NP_808403.2 | 3 | 562 | ||
| GenBank mRNA | BC067004.1 | - | 3 | 562 | 6416222 | |
| BC127616.1 | - | 3 | 539 | 40093666 | ||
| BC127617.1 | - | 3 | 539 | 40093667 | ||

Tmem130
Genomic Map