siRNA Details
Gene Symbol: Tmem130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177735.4 | NP_808403.2 | 1, 2 | 127 | ||
GenBank mRNA | BC067004.1 | - | 1, 2 | 127 | 6416222 | |
BC127616.1 | - | 1, 2 | 104 | 40093666 | ||
BC127617.1 | - | 1, 2 | 104 | 40093667 |
