siRNA Details
| Gene Symbol: Gprin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183183.2 | NP_899006.1 | 2 | 1897 | ||
| GenBank mRNA | AK050148.1 | - | 2 | 1898 | ||
| AK173336.1 | - | 2 | 1901 | |||
| BC138149.1 | - | 1 | 1767 | 8861168 | ||
| BC138150.1 | - | 1 | 1767 | 8861169 | ||

Gprin3
Genomic Map