siRNA Details
| Gene Symbol: Ccdc142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081266.1 | NP_001074735.1 | 3 | 1263 | ||
| GenBank mRNA | AK038734.1 | - | 1 | 1516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc142| Gene Symbol: Ccdc142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081266.1 | NP_001074735.1 | 3 | 1263 | ||
| GenBank mRNA | AK038734.1 | - | 1 | 1516 | ||