siRNA Details
| Gene Symbol: Kbtbd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008785.5 | NP_001008785.2 | 3 | 528 | ||
| NM_001102670.2 | NP_001096140.1 | 2 | 317 | |||
| GenBank mRNA | AK142080.1 | - | 3 | 502 | ||
| AK167507.1 | - | 2 | 268 | |||
| BC088734.1 | - | 2 | 439 | 30459462 | ||

Kbtbd8
Genomic Map