siRNA Details
| Gene Symbol: Slc6a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172890.3 | NP_766478.1 | 12 | 1513 | ||
| GenBank mRNA | AK053078.1 | - | 12 | 1514 | ||
| AK053261.1 | - | 12 | 1355 | |||
| AK140423.1 | - | 12 | 1516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a11| Gene Symbol: Slc6a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172890.3 | NP_766478.1 | 12 | 1513 | ||
| GenBank mRNA | AK053078.1 | - | 12 | 1514 | ||
| AK053261.1 | - | 12 | 1355 | |||
| AK140423.1 | - | 12 | 1516 | |||