siRNA Details
| Gene Symbol: Ppp1r9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181595.3 | NP_853626.1 | 2 | 592 | ||
| GenBank mRNA | AB091828.1 | - | 2 | 592 | ||
| AK030399.1 | - | 2 | 581 | |||
| AK135170.1 | - | 2 | 492 | |||
| AK138313.1 | - | 2 | 521 | |||
| AK158559.1 | - | 2 | 540 | |||
| AY308065.1 | - | 1 | 267 | |||
| BC053748.1 | - | 2 | 480 | 6331919 | ||

Ppp1r9a
Genomic Map