siRNA Details
| Gene Symbol: Plxna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175750.3 | NP_786926.2 | 10 | 3185 | ||
| GenBank mRNA | AB073228.1 | - | 10 | 3184 | ||
| AK051217.1 | - | 1 | 1174 | |||
| BC141298.1 | - | 10 | 2483 | 9055910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna4| Gene Symbol: Plxna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175750.3 | NP_786926.2 | 10 | 3185 | ||
| GenBank mRNA | AB073228.1 | - | 10 | 3184 | ||
| AK051217.1 | - | 1 | 1174 | |||
| BC141298.1 | - | 10 | 2483 | 9055910 | ||