siRNA Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172892.3 | NP_766480.1 | 3 | 910 | ||
| GenBank mRNA | AK036354.1 | - | 3 | 912 | ||
| AK038937.1 | - | 3 | 915 | |||
| AK139428.1 | - | 3 | 913 | |||
| BC089161.1 | - | 3 | 899 | 30440173 | ||

Slc13a4
Genomic Map