siRNA Details
| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203491.3 | NP_987076.2 | 3 | 799 | ||
| GenBank mRNA | BC019470.1 | - | 3 | 799 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrm2| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203491.3 | NP_987076.2 | 3 | 799 | ||
| GenBank mRNA | BC019470.1 | - | 3 | 799 | ||
Set Membership: |
|