siRNA Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172893.3 | NP_766481.2 | 4 | 1048 | ||
| GenBank mRNA | AK036886.1 | - | 4 | 1051 | ||
| AK053205.1 | - | 4 | 949 | |||
| AK081528.1 | - | 4 | 949 | |||
| AK156623.1 | - | 4 | 917 | |||
| AK162200.1 | - | 4 | 619 | |||
| BC120733.1 | - | 4 | 881 | 40129656 | ||
| BC137645.1 | - | 4 | 881 | 8860664 | ||

Parp12
Genomic Map