siRNA Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172893.3 | NP_766481.2 | 11 | 1911 | ||
| GenBank mRNA | AK036886.1 | - | 11 | 1914 | ||
| AK053205.1 | - | 11 | 1812 | |||
| AK081528.1 | - | 11 | 1813 | |||
| AK156623.1 | - | 11 | 1780 | |||
| AK162200.1 | - | 11 | 1483 | |||
| BC024579.1 | - | 6 | 560 | |||
| BC028906.1 | - | 3 | 270 | |||
| BC048927.1 | - | 7 | 762 | |||
| BC120733.1 | - | 11 | 1744 | 40129656 | ||
| BC137645.1 | - | 11 | 1744 | 8860664 | ||

Parp12
Genomic Map