siRNA Details
| Gene Symbol: Zfp324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178732.3 | NP_848847.3 | 4 | 1524 | ||
| GenBank mRNA | AK044240.1 | - | 4 | 1521 | ||
| AK051488.1 | - | 4 | 1547 | |||
| AK052251.1 | - | 3 | 1417 | |||
| AK054407.1 | - | 4 | 1569 | |||
| AK085053.1 | - | 3 | 1420 | |||
| AK152107.1 | - | 4 | 1550 | |||
| AK152696.1 | - | 4 | 1467 | |||
| AK165922.1 | - | 4 | 1520 | |||
| BC053093.1 | - | 4 | 1524 | 6400158 | ||

Zfp324
Genomic Map