siRNA Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172898.3 | NP_766486.1 | 15 | 2559 | ||
| GenBank mRNA | AK049284.1 | - | 15 | 2560 | ||
| AK083069.1 | - | 13 | 3109 | |||
| BC052773.1 | - | 15 | 2022 | 6314924 | ||
| BC158021.1 | - | 15 | 2067 | 9007327 | ||

Kirrel2
Genomic Map