siRNA Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172898.3 | NP_766486.1 | 5 | 1318 | ||
| GenBank mRNA | AK049284.1 | - | 5 | 1319 | ||
| AK083069.1 | - | 5 | 1461 | |||
| AY305303.1 | - | 4 | 456 | |||
| BC052773.1 | - | 5 | 781 | 6314924 | ||
| BC158021.1 | - | 5 | 826 | 9007327 | ||

Kirrel2
Genomic Map