siRNA Details
| Gene Symbol: 4930433I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207248.3 | NP_997131.2 | 2 | 1083 | ||
| GenBank mRNA | AK029662.1 | - | 2 | 1139 | ||
| BC150916.1 | - | 2 | 1083 | 9087827 | ||
| BC150921.1 | - | 2 | 1083 | 9087832 | ||

4930433I11Rik
Genomic Map