siRNA Details
| Gene Symbol: Shank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034115.1 | NP_001029287.1 | 24 | 4778 | ||
| GenBank mRNA | AK082624.1 | - | 2 | 2152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shank1| Gene Symbol: Shank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034115.1 | NP_001029287.1 | 24 | 4778 | ||
| GenBank mRNA | AK082624.1 | - | 2 | 2152 | ||