siRNA Details
| Gene Symbol: Ntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033356.3 | NP_001028528.2 | 3 | 1160 | ||
| NM_001289692.1 | NP_001276621.1 | 3 | 1160 | |||
| NM_001289693.1 | NP_001276622.1 | 4 | 953 | |||
| GenBank mRNA | AK042348.1 | - | 4 | 955 | ||
| AK142898.1 | - | 3 | 768 | |||
| BC150901.1 | - | 3 | 753 | 9087812 | ||
| BC172055.1 | - | 3 | 753 | |||

Ntn5
Genomic Map