siRNA Details
| Gene Symbol: Ntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033356.3 | NP_001028528.2 | 2 | 556 | ||
| NM_001289692.1 | NP_001276621.1 | 2 | 556 | |||
| GenBank mRNA | AK142898.1 | - | 2 | 164 | ||
| BC150901.1 | - | 2 | 149 | 9087812 | ||
| BC172055.1 | - | 2 | 149 | |||

Ntn5
Genomic Map