siRNA Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028031.3 | NP_082307.1 | 7, 8 | 848 | ||
| GenBank mRNA | AK010382.1 | - | 7, 8 | 848 | ||
| AK049640.1 | - | 6, 7 | 678 | |||
| AK145855.1 | - | 7, 8 | 768 | |||
| AK146196.1 | - | 7, 8 | 820 | |||
| BC019493.1 | - | 7, 8 | 767 | |||
| BC046599.1 | - | 7, 8 | 767 | 6311721 | ||

Zdhhc13
Genomic Map