siRNA Details
| Gene Symbol: Fsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172904.2 | NP_766492.2 | 2 | 329 | ||
| GenBank mRNA | AK028142.1 | - | 2 | 321 | ||
| AK036674.1 | - | 295 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fsd2| Gene Symbol: Fsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172904.2 | NP_766492.2 | 2 | 329 | ||
| GenBank mRNA | AK028142.1 | - | 2 | 321 | ||
| AK036674.1 | - | 295 | ||||